Glass - trace elements, NIST standards
NIST-613 glass - trace elements, 1
Catalog # PROONIST-613
Supplier: LGC Standards PROMOCHEM
Glass - trace elements, NIST standards
Catalog # PROONIST-613
Supplier: LGC Standards PROMOCHEM
CAS Number:
Jump to:
Product Details & Documents
This standard reference material is intended to facilitate the development of chemical methods of analysis for trace elements.
Certifications: Traceable to NIST and SRM.
Specifications
Specifications
- Pk:1
- Description:NIST-613 glass - trace elements