Orders LinkContactUsLinkComponent
Glass - trace elements, NIST standards
Glass - trace elements, NIST standards
NIST-612 glass - trace elements, 1
Catalog # PROONIST-612
Glass - trace elements, NIST standards
Catalog # PROONIST-612
CAS Number:  
Glass - trace elements, NIST standards
NIST-612 glass - trace elements, 1
CAS Number:  
Price
1 110,00 €
Log in for availability
1 * 1 items
Product Details & Documents
This standard reference material is intended to facilitate the development of chemical methods of analysis for trace elements.
Certifications: Traceable to NIST and SRM.
Specifications
Specifications
  • Pk:
    1
  • Description:
    NIST-612 glass - trace elements
Specifications