Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SUC-LEU-LEU-VAL-TYR-7-AMINO-4-METHYLCOUM 1 * 25 mg
Catalog # ICNA03AMC08625
Supplier:
MP Biomedicals
Expand image
SUC-LEU-LEU-VAL-TYR-7-AMINO-4-METHYLCOUM 1 * 25 mg
Catalog # ICNA03AMC08625
Supplier:
MP Biomedicals