Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
LYSING MATRIX A 1 * 500 items
LYSING MATRIX A 1 * 500 items
Print
LYSING MATRIX A 1 * 500 items
Catalog # ICNA6910500
Supplier:
MP Biomedicals
LYSING MATRIX A 1 * 500 items
Catalog # ICNA6910500
Supplier:
MP Biomedicals
Expand image
Return to Top
LYSING MATRIX A 1 * 500 items
Add to cart
Price
1 472,00 €
Log in
for availability
Unit of Measure
1 * 500 items
-
+
Add to cart
Return Policy
Product Details & Documents