0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
N-DECANE 1 * 6 L
N-DECANE 1 * 6 L
ICNA0230005106
:
MP Biomedicals
N-DECANE 1 * 6 L
ICNA0230005106
:
MP Biomedicals
:
N-DECANE 1 * 6 L
:
734,00 €
1 * 6 L
-
+