G418 disulphate in solution
Glass bottle, 20 mL
A6798.0020
G418 disulphate supplied as a ready-to-use sterile solution.
Concentration: 50 mg/ml
: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
- :108321-42-2
- :C₂₀H₄₄N₄O₁₈S₂
- :MFCD00058314
- :692.72 g/mol
- Pack type:Glass bottle
- Pk:20 mL