Order Entry
Export
Orders LinkContactUsLinkComponent
Zinc fluoride 98%

Zinc fluoride 98%

Plastic bottle, 250 g
Catalog # ACRO363912500
Corrosive
Irritant
Zinc fluoride 98%
Catalog # ACRO363912500
CAS Number:  7783-49-5
Zinc fluoride 98%
Zinc fluoride 98%

Plastic bottle, 250 g

CAS Number:  7783-49-5
Price
294,00 €
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:7783-49-5
  • Formula:F₂Zn
  • MDL Number:MFCD00011298
  • Molecular Weight:103.39 g/mol
  • Boiling Pt:1500 °C (1013 hPa)
  • Melting Pt:872…910 °C
  • Density:4,95 g/cm³ (20 °C)
  • EINECS:232-001-9
  • ADR:6.1,III
  •  :Density: 4.95 g/cm³ (20 °C)

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Fluoride (F):
    36.0 to 37.8 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g

Specifications