Zinc fluoride 98%
Plastic bottle, 25 g
- :7783-49-5
- :F₂Zn
- :MFCD00011298
- :103.39 g/mol
- :1500 °C (1013 hPa)
- :872…910 °C
- :4,95 g/cm³ (20 °C)
- :232-001-9
- :6.1,III
- :Density: 4.95 g/cm³ (20 °C)
- Appearance:White to off-white Powder
- Fluoride (F):36.0 to 37.8 %
- Pack type:Plastic bottle
- Pk:25 g



