Ethyl acetate 99.5% for analysis
Metal drum, 25 L
Ethyl acetate 99.5% for analysis
232110250.
: 141-78-6
: Acetic acid ethyl ester
- :141-78-6
- :CH₃COOC₂H₅
- :MFCD00009171
- :88.11 g/mol
- :13,03792
- :75…78 °C (760 mmHg)
- :–83.5 °C
- :0.902 g/cm³
- :–4 °C
- :Ambient
- :205-500-4
- :1173
- :3,II
- :01-2119475103-46
- Appearance:Clear liquid
- Water:≤0.05 %
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0005 meq/g corresp. 0.003% HOAc
- Substance darkened by H2SO4:≤10 APHA
- Aluminium (Al):≤0.5 ppm
- Barium (Ba):≤0.1 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- Ethanol:≤0.1 %
- Methanol:≤0.1 %
- Methyl acetate:≤0.1 %
- Pack type:Metal drum
- Pk:25 L
- Cat. No.:232110250.
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health



