Ethyl acetate 99.5% for analysis
Plastic drum, 5 L
Supplier: Thermo Fisher Scientific
Ethyl acetate 99.5% for analysis
Catalog # 232110050.
Supplier: Thermo Fisher Scientific
CAS Number: 141-78-6
Jump to:
Product Details & Documents
Synonyms: Acetic acid ethyl ester
- CAS Number:141-78-6
- Formula:CH₃COOC₂H₅
- MDL Number:MFCD00009171
- Molecular Weight:88.11 g/mol
- Merck Index:13,03792
- Boiling Pt:75…78 °C (760 mmHg)
- Melting Pt:–83.5 °C
- Density:0.902 g/cm³
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- EINECS:205-500-4
- UN:1173
- ADR:3,II
- REACH:01-2119475103-46
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.05 %
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0005 meq/g corresp. 0.003% HOAc
- Substance darkened by H2SO4:≤10 APHA
- Aluminium (Al):≤0.5 ppm
- Barium (Ba):≤0.1 ppm
- Calcium (Ca):≤0.5 ppm
- Chromium (Cr):≤0.02 ppm
- Cobalt (Co):≤0.02 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Zinc (Zn):≤0.1 ppm
- Ethanol:≤0.1 %
- Methanol:≤0.1 %
- Methyl acetate:≤0.1 %
Specifications
- Pack type:Plastic drum
- Pk:5 L
- Cat. No.:232110050.
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health



