Order Entry
Export
Orders LinkContactUsLinkComponent
 

Chloroform 99+% stabilised for analysis

Metal drum, 25 L
Catalog # ACRO232090250
Toxic
Health
Chloroform 99+% stabilised for analysis
Catalog # ACRO232090250
CAS Number:  67-66-3
Chloroform 99+% stabilised for analysis
Chloroform 99+% stabilised for analysis

Metal drum, 25 L

CAS Number:  67-66-3
Price
1 260,00 €
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61 °C (760 mmHg)
  • Melting Pt:–63 °C
  • Density:1.48 g/cm³
  • EINECS:200-663-8
  • ADR:6.1,III
  •  :Stabilised with ethanol
  • Storage Temperature:Ambient
  • UN:1888
  • REACH:05-2114614906-45
Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.01 %
  • GC:
    ≥99.0 % (Excluding stabilizer)
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    0.6 to 1.0 % ethanol
  • Residue after evaporation:
    ≤0.001 %
  • Free chlorine:
    Passes test
  • Cadmium (Cd):
    ≤0.1 ppm
  • Cobalt (Co):
    ≤0.1 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.5 ppm
  • Nickel (Ni):
    ≤0.1 ppm
  • Zinc (Zn):
    ≤0.5 ppm
  • Acidity (HCl):
    ≤0.001 %
  • Acetone and aldehyde:
    ≤0.005 % (as Acetone)

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 L

Specifications