Order Entry
Export
Orders LinkContactUsLinkComponent
1-Tetradecanol 99%

1-Tetradecanol 99%

Glass bottle for solids, 5 g
Catalog # ACRO180730050
Irritant
Environment
1-Tetradecanol 99%
Catalog # ACRO180730050
CAS Number:  112-72-1
1-Tetradecanol 99%
1-Tetradecanol 99%

Glass bottle for solids, 5 g

CAS Number:  112-72-1
Price
29,60 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol
  • CAS Number:112-72-1
  • Formula:H₃C(CH₂)₁₂CH₂OH
  • MDL Number:MFCD00004757
  • Molecular Weight:214.39 g/mol
  • Merck Index:13,06361
  • Boiling Pt:289 °C (760 mmHg)
  • Melting Pt:37…40 °C
  • Density:0,8 g/cm³ (60 °C)
  • Flash Pt:145 °C
  • Storage Temperature:Ambient
  • EINECS:204-000-3
  • UN:3077
  • ADR:9,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals or flakes or pellets and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    37 to 41 °C
  • GC:
    ≥98.5 %
  • Titration with HClO4:
    97.5 to 102.5 % (on dry substance)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications