2-Chlorophenylboronic acid 97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
2-Chlorophenylboronic acid 97%
Catalog # ACRO358990050
Supplier: Thermo Fisher Scientific
CAS Number: 3900-89-8
Jump to:
Product Details & Documents
Synonyms: (2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid
- CAS Number:3900-89-8
- Formula:C₆H₆BClO₂
- MDL Number:MFCD00674012
- Molecular Weight:156.38 g/mol
- Melting Pt:94…95 °C
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:White to light beige Powder or crystals or granules
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



