Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,4-Dichloropyridine 98%

Glass bottle for solids, 5 g
Catalog # ACRO379470050
Irritant
2,4-Dichloropyridine 98%
Catalog # ACRO379470050
CAS Number:  26452-80-2
2,4-Dichloropyridine 98%
2,4-Dichloropyridine 98%

Glass bottle for solids, 5 g

CAS Number:  26452-80-2
Price
89,40 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:26452-80-2
  • Formula:C₅H₃Cl₂N
  • MDL Number:MFCD00955618
  • Molecular Weight:147.99 g/mol
  • Melting Pt:–1 °C
  • Density:1.37 g/cm³
  • Flash Pt:89 °C
  • Storage Temperature:Ambient
  • EINECS:247-717-7
  • ADR:6.1,III

 

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5500 to 1.5540 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications