2,7-Dibromofluorene 99%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
2,7-Dibromofluorene 99%
Catalog # ACRO355240050
Supplier: Thermo Fisher Scientific
CAS Number: 16433-88-8
Jump to:
Product Details & Documents
Synonyms: 2,7-Dibromo-9H-fluorene
- CAS Number:16433-88-8
- Formula:C₁₃H₈Br₂
- MDL Number:MFCD00019048
- Molecular Weight:324.01 g/mol
- Melting Pt:164…167 °C
- Storage Temperature:Ambient
Documents
ACRO35524
SDS
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:164 to 167 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



