Order Entry
Export
Orders LinkContactUsLinkComponent
 

Hydrocortisone 98%

Glass bottle, 1 g
Catalog # ACRO352450010
Health
Hydrocortisone 98%
Catalog # ACRO352450010
CAS Number:  50-23-7
Hydrocortisone 98%
Hydrocortisone 98%

Glass bottle, 1 g

CAS Number:  50-23-7
Price
39,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:50-23-7
  • Formula:C₂₁H₃₀O₅
  • MDL Number:MFCD00011654
  • Molecular Weight:362.47 g/mol
  • Boiling Pt:567 °C (1013 hPa)
  • Melting Pt:212…217 °C
  • Density:1,4 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-020-1

 

Specifications

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    212 to 217 °C
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +150° to +156° (20 °C, 589 nm) (C=1, dioxane)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications