Triethyl citrate 99%
Plastic bottle, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Triethyl 2-hydroxypropane-1,2,3-tricarboxylate, Hydroxytricarballylic acid triethyl ester, Citric acid triethyl ester
- CAS Number:77-93-0
- Formula:C₁₂H₂₀O₇
- MDL Number:MFCD00009201
- Molecular Weight:276.29 g/mol
- Boiling Pt:294 °C (760 mmHg)
- Melting Pt:–46 °C
- Density:1.136 g/cm³
- Flash Pt:151 °C
- Storage Temperature:Ambient
- EINECS:201-070-7
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4400 to 1.4430 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤50 APHA
Specifications
- Pack type:Plastic bottle
- Pk:250 g



