Order Entry
Export
Orders LinkContactUsLinkComponent
n-Pentane for analysis

n-Pentane for analysis

Glass bottle, 2,5 L
Catalog # ACRO383900025
Irritant
Health
Environment
Flammable
n-Pentane for analysis
Catalog # ACRO383900025
CAS Number:  109-66-0
n-Pentane for analysis
n-Pentane for analysis

Glass bottle, 2,5 L

CAS Number:  109-66-0
Price
200,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Pentane
  • CAS Number:109-66-0
  • Formula:H₃C(CH₂)₃CH₃
  • MDL Number:MFCD00009498
  • Molecular Weight:72.15 g/mol
  • Merck Index:13,07193
  • Boiling Pt:36 °C (760 mmHg)
  • Melting Pt:–130 °C
  • Density:0.626 g/cm³
  • Flash Pt:–49 °C
  • Storage Temperature:Ambient
  • EINECS:203-692-4
  • UN:1265
  • ADR:3,II
  • REACH:01-2119459286-30

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.01 % (K.F.)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.002 %
  • Acidity/alkalinity:
    ≤0.0001 meq/g
  • Calcium (Ca):
    ≤0.5 ppm
  • Copper (Cu):
    ≤0.05 ppm
  • Iron (Fe):
    ≤0.2 ppm
  • Lead (Pb):
    ≤0.05 ppm
  • Magnesium (Mg):
    ≤0.1 ppm
  • Potassium (K):
    ≤0.2 ppm
  • Sodium (Na):
    ≤1 ppm
  • Zinc (Zn):
    ≤0.1 ppm
  • Total phosphorus (P):
    ≤0.5 ppm
  • Total silicon (Si):
    ≤0.02 ppm
  • Total sulfur (S):
    ≤5 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications