Chloroform 99.8% stabilised for analysis
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Chloroform 99.8% stabilised for analysis
Catalog # ACRO383770025
Supplier: Thermo Fisher Scientific
CAS Number: 67-66-3
Jump to:
Product Details & Documents
Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
- CAS Number:67-66-3
- Formula:CHCl₃
- MDL Number:MFCD00000826
- Molecular Weight:119.38 g/mol
- Merck Index:13,02160
- Boiling Pt:61 °C (760 mmHg)
- Melting Pt:–63 °C
- Density:1.48 g/cm³
- EINECS:200-663-8
- ADR:6.1,III
- :Stabilised with amylene
- Storage Temperature:Ambient
- UN:1888
- REACH:05-2114614906-45
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.02 % (K.F.)
- GC:≥99.8 %
- Color scale:≤5 APHA
- Stabilizer:Approx. 100 ppm Amylene
- Residue after evaporation:≤5 ppm
- Acidity/alkalinity:≤0.0005 meq/g
- Suitability:(for use with Dithizone) Passes test
- Substance darkened by H2SO4:≤5 APHA
- Free chlorine:≤5 ppm
- Calcium (Ca):≤0.1 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.02 ppm
- Magnesium (Mg):≤0.05 ppm
- Potassium (K):≤0.1 ppm
- Sodium (Na):≤0.2 ppm
- Zinc (Zn):≤0.05 ppm
- Impurity:≤0.005 % Carbon tetrachloride
- Total phosphorus (P):≤0.1 ppm
- Total silicon (Si):≤0.05 ppm
- Total sulfur (S):≤0.5 ppm
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



