Zinc stearate
Plastic bottle, 1 kg
: Stearic acid zinc salt, Zinc(II) stearate
- :557-05-1
- :C₃₆H₇₀O₄Zn
- :MFCD00013031
- :632.34 g/mol
- :13,10212
- :118…128 °C
- :1,095 g/cm³ (20 °C)
- :>100 °C
- :Ambient
- :209-151-9
- Appearance:White Powder
- Infrared spectrum:Conforms
- Titration Complexometric:12.5 to 14.0 % (ZnO)
- Pack type:Plastic bottle
- Pk:1 kg



