1,9-Dibromononane 97%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
1,9-Dibromononane 97%
Catalog # ACRO112870250
Supplier: Thermo Fisher Scientific
CAS Number: 4549-33-1
Jump to:
Product Details & Documents
Synonyms: Nonamethylene dibromide
- CAS Number:4549-33-1
- Formula:BrH₂C(CH₂)₇CH₂Br
- MDL Number:MFCD00000279
- Molecular Weight:286.05 g/mol
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:–10 °C
- Density:1.40 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:224-913-0
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4950 to 1.4980 (20°C, 589 nm)
- GC:≥96.0 %
- Specific optical rotation:+39° to +69° (20°C, 589 nm) (C=2, Chloroform)
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Cat. No.:ACRO112870250



