Order Entry
Export
Orders LinkContactUsLinkComponent
1,9-Dibromononane 97%

1,9-Dibromononane 97%

Glass bottle for solids, 5 g
Catalog # ACRO112870050
Irritant
1,9-Dibromononane 97%
Catalog # ACRO112870050
CAS Number:  4549-33-1
1,9-Dibromononane 97%
1,9-Dibromononane 97%

Glass bottle for solids, 5 g

CAS Number:  4549-33-1
Price
31,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Nonamethylene dibromide
  • CAS Number:4549-33-1
  • Formula:BrH₂C(CH₂)₇CH₂Br
  • MDL Number:MFCD00000279
  • Molecular Weight:286.05 g/mol
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:–10 °C
  • Density:1.40 g/cm³
  • Flash Pt:>112 °C
  • Storage Temperature:Ambient
  • EINECS:224-913-0

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4950 to 1.4980 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Specific optical rotation:
    +39° to +69° (20°C, 589 nm) (C=2, Chloroform)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications