3,5-Dinitroaniline 98%
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
3,5-Dinitroaniline 98%
Catalog # ACRO116940100
Supplier: Thermo Fisher Scientific
CAS Number: 618-87-1
Jump to:
Product Details & Documents
- CAS Number:618-87-1
- Formula:C₆H₅N₃O₄
- MDL Number:MFCD00007263
- Molecular Weight:183.12 g/mol
- Melting Pt:159…163 °C
- Density:1,601 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:210-567-8
- UN:1596
- ADR:6.1,II
Documents
ACRO11694
SDS
Specifications
Specification Test Results
- Appearance:Yellow to light brown Powder
- Infrared spectrum:Conforms
- Melting point:159 to 163 °C
- HPLC:≥97.5 %
- Calcium (Ca):≤50 ppm
- Copper (Cu):≤20 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤20 ppm
- Magnesium (Mg):≤50 ppm
- Nickel (Ni):≤1500 ppm
- Potassium (K):≤100 ppm
- Sodium (Na):≤500 ppm
- Zinc (Zn):≤100 ppm
- Insoluble matter:≤0.01 %
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



