Order Entry
Export
Orders LinkContactUsLinkComponent
 

Benzophenone imine 97% stabilised

Glass bottle for solids, 25 g
Catalog # ACRO368590250
Irritant
Benzophenone imine 97% stabilised
Catalog # ACRO368590250
CAS Number:  1013-88-3
Benzophenone imine 97% stabilised
Benzophenone imine 97% stabilised

Glass bottle for solids, 25 g

CAS Number:  1013-88-3
Price
264,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Benzophenoneimine
  • CAS Number:1013-88-3
  • Formula:C₁₃H₁₁N
  • MDL Number:MFCD00001760
  • Molecular Weight:181.24 g/mol
  • Boiling Pt:216 °C (75 mmHg)
  • Melting Pt:–30 °C
  • Density:1.08 g/cm³
  • Flash Pt:145 °C
  • Storage Temperature:Ambient
  • ADR:9,III

 

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow-brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6170 to 1.6190 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Stabilizer:
    Approx. 0.02 % w/w Hydroquinone

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications