2-Methylresorcinol 98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
2-Methylresorcinol 98%
Catalog # ACRO364740250
Supplier: Thermo Fisher Scientific
CAS Number: 608-25-3
Jump to:
Product Details & Documents
Synonyms: 2,6-Dihydroxytoluene, 2-Methylbenzene-1,3-diol
- CAS Number:608-25-3
- Formula:C₇H₈O₂
- MDL Number:MFCD00002271
- Molecular Weight:124.14 g/mol
- Boiling Pt:264 °C (1013 hPa)
- Melting Pt:118…124 °C
- Flash Pt:135 °C
- Storage Temperature:Ambient
- EINECS:210-155-8
- UN:2811
- ADR:6.1,III
Documents
ACRO36474
SDS
Specifications
Specification Test Results
- Appearance:Beige to pink to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:118 to 124 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



