Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Media for Microbiology
Additives and Supplements for Microbiology
Coagulase plasma
Coagulase plasma
Print
Coagulase plasma
Coagulase Plasma lyophilised, 5 mL
Catalog # OXOIR21050
Supplier:
THERMO FISHER DIAGNOSTICS
Coagulase plasma
Catalog # OXOIR21050
Supplier:
THERMO FISHER DIAGNOSTICS
CAS Number:
Expand image
Return to Top
Coagulase plasma
Coagulase Plasma lyophilised, 5 mL
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
71,00 €
Log in
for availability
Unit of Measure
1 * 5 mL
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Specifications
Pk:
5 mL
Description:
Coagulase Plasma lyophilised
Specifications