Order Entry
Export
Orders LinkContactUsLinkComponent
Contact plates

Contact plates

Sabouraud glucose Chloramphenicol agar, BP/EP/JP/USP, 10
Catalog # OXOIPO5094C
Contact plates
Catalog # OXOIPO5094C
CAS Number:  
Contact plates

Sabouraud glucose Chloramphenicol agar, BP/EP/JP/USP, 10

CAS Number:  
Price
32,70 €
Log in for availability
1 * 10 items
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    10
  • Standards:
    BP/EP/JP/USP
  • Description:
    Sabouraud glucose Chloramphenicol agar

Specifications