Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Media for Microbiology
Contact Plates for Microbiology
Contact plates
Contact plates
Print
Contact plates
Malt extract agar, 10
Catalog # OXOIPO5079C
Supplier:
THERMO FISHER DIAGNOSTICS
Contact plates
Catalog # OXOIPO5079C
Supplier:
THERMO FISHER DIAGNOSTICS
CAS Number:
Expand image
Return to Top
Contact plates
Malt extract agar, 10
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
29,40 €
Log in
for availability
Unit of Measure
1 * 10 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
Datasheet
OXOIPO5094C_TDS_EN
Specifications
Specifications
Pk:
10
Description:
Malt extract agar
Specifications