Inclusion bodies solubilisation buffer
IBS™ buffer, 100 mL
Catalog # 786-183
Supplier: G-Biosciences
Jump to:
Product Details & Documents
The IBS™ buffer is specifically developed for solubilisation of inclusion bodies.
IBS-HP™ Buffer is for the solubilisation of inclusion bodies containing highly hydrophobic proteins.
Specifications
Specifications
- Pk:100 mL
- Description:IBS™ buffer