Order Entry
Export
Orders LinkContactUsLinkComponent
 

Agarose coupling kits, HOOK™

HOOK™ Activated Agarose (amine reactive), 10 mL
Catalog # 786-066
Supplier:   G-Biosciences
Agarose coupling kits, HOOK™
Catalog # 786-066
Supplier:  G-Biosciences
CAS Number:  
Agarose coupling kits, HOOK™
Agarose coupling kits, HOOK™

HOOK™ Activated Agarose (amine reactive), 10 mL

CAS Number:  
Price
226,00 €
Log in for availability
1 * 10 mL
Product Details & Documents

The amine reactive HOOK™ Agarose Coupling Kit is designed for the simple and efficient coupling of proteins and peptides to a solid support. The kit allows the generation of both a stable and reusable column for the affinity purification of antibodies, proteins and other protein interacting biomolecules.

This kit utilises our HOOK™ Activated Agarose (Amine) that has reactive aldehyde groups. The aldehyde groups of the agarose react spontaneously with primary amines, located at the N-terminus of proteins or in lysine residues, to form intermediate Schiff Base complexes. These, in turn, are selectively reduced by the supplied reduction agent. Following quenching of the reaction the protein or peptide is linked to the agarose by stable amine linkage. The resin can now be aliquoted into the supplied columns for use in purification and interation studies.

In addition to all the coupling reagents, the HOOK™ Agarose Coupling Kit is supplied with a specific Optimizer Buffer™ that provides the optimal coupling conditions and can be used to equilibrate your starting protein sample.

This kit is suitable for the preparation of 5 stable columns containing 2 ml resin.

Specifications

Specifications

  • Pk:
    10 mL
  • Description:
    HOOK™ Activated Agarose (amine reactive)

Specifications