Order Entry
Export
Orders LinkContactUsLinkComponent
TE Buffers 100X

TE Buffers 100X

TE buffer, 100X, 8,0, Plastic bottle, 100 mL
Catalog # R003
Supplier:   G-Biosciences
TE Buffers 100X
Catalog # R003
Supplier:  G-Biosciences
CAS Number:  
TE Buffers 100X

TE buffer, 100X, 8,0, Plastic bottle, 100 mL

CAS Number:  
Price
45,80 €
Log in for availability
1 * 100 mL
Product Details & Documents

TE Buffer (Tris-EDTA Buffer) is a common molecular biology buffer that is used to protect DNA and RNA from degradation. The Tris buffering agent and EDTA metal chelating properties help protect DNA and RNA.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    TE buffer, 100X
  • pH value:
    8,0

Specifications