Order Entry
Export
Orders LinkContactUsLinkComponent
ASB-16 ≥99%

ASB-16 ≥99%

5 g
Catalog # DG063
Supplier:   G-Biosciences
ASB-16 ≥99%
Catalog # DG063
Supplier:  G-Biosciences
CAS Number:  52562-29-5
ASB-16 ≥99%

5 g

CAS Number:  52562-29-5
Price
879,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • Zwitterionic detergent
  • Aminosulfobetaine with C16 alkyl tail
  • In some cases superior than ASB-14
  • Useful for solubilizing proteins for 2D analysis
  • Optimal solubility achieved in urea-thiourea mixtures and not in urea alone
  • Reported to show better protein solubilization properties than CHAPS
  • ASB-16 has been shown to solubilize membrane proteins previously undetected
Synonyms:  Amidosulphobetaine-16
  • CAS Number:52562-29-5
  • Formula:C₂₄H₅₀N₂O₄S
  • Molecular Weight:462.734 g/mol
Specifications

Specifications

  • Pk:
    5 g

Specifications