Biotin conjugation estimation, HOOK™ BiotinQuant
HABA dye, 1 g
Catalog # BKC-03
Supplier: G-Biosciences
Biotin conjugation estimation, HOOK™ BiotinQuant
Catalog # BKC-03
Supplier: G-Biosciences
CAS Number:
Jump to:
Product Details & Documents
HOOK™ BiotinQuant is designed for the estimation of biotin conjugation. It measures biotin using HABA [4’-hydroxyazobenzene-2-carboxylic acid] dye. HABA binds with avidin at the biotin-binding site.
Documents
BKC-01_protocol
Manual
Specifications
Specifications
- Pk:1 g
- Description:HABA dye