Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
25 L
Supplier: Avantor
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 8254.9025
Supplier: Avantor
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Boiling Pt:35 °C (1013 hPa)
- Melting Pt:–123 °C
- Density:0.7135 g/cm³ (20 °C)
- Flash Pt:–40 °C
- Storage Temperature:Ambient
- EINECS:200-467-2
- UN:1155
- ADR:3,I
- REACH:01-2119535785-29
Documents
Specifications
Specification Test Results
- Assay (by GC):min. 99,5 %
- Alcohol (C2H5OH):max. 0,01 %
- Carbonyl Compounds (as HCHO):max. 0,001 %
- Color (APHA):max. 10
- Density (g/ml) at 20°C:0.712-0.714
- Foreign Odor:passes test
- Peroxide (as H202):max. 3 ppm
- Residue after Evaporation:max. 0,001 %
- Substances Darkened by H2SO4:passes test
- Titrable Acid (meq/g):max. 0,0002
- Water (H2O):max. 0,01 %
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,02
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
Specifications
- Pk:25 L



