Orders LinkContactUsLinkComponent
Funnels, powder
80 mm
Catalog # WITG5.790.004
Supplier:   witeg Labortechnik
Funnels, powder
Catalog # WITG5.790.004
Supplier:  witeg Labortechnik
Funnels, powder
Funnels, powder
80 mm
Price
17,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Powder funnels, made of DURAN® borosilicate glass 3.3, with short wide stem.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Funnel O-Ø:
    80 mm