Adapters, delivery, straight
24/29 NS, 100 mm
Catalog # WITG1.900.024
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
- Socket size:24/29 NS
- Tube length:100 mm