Orders LinkContactUsLinkComponent
Immersion coolers
Immersion coolers
100 mm, 19/26 NS
Catalog # WITG1.786.219
Supplier:   witeg Labortechnik
Immersion coolers
Catalog # WITG1.786.219
Supplier:  witeg Labortechnik
Immersion coolers
Immersion coolers
100 mm, 19/26 NS
Price
53,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For immersion into distillation assemblies etc. Can be used to regulate refluxing by adjusting the flow of air or water through it. Water connection with two glass olives.
Specifications
  • Cone size:
    19/26 NS
  • Jacket length:
    100 mm