Orders LinkContactUsLinkComponent
Distilling heads
Distilling heads
24/29 NS, 24/29 NS
Catalog # WITG1.010.005
Supplier:   witeg Labortechnik
Distilling heads
Catalog # WITG1.010.005
Supplier:  witeg Labortechnik
Distilling heads
Distilling heads
24/29 NS, 24/29 NS
Price
34,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With one NS socket and two NS cones, descending angle: 75°.
Specifications
  • Cone to condenser:
    24/29 NS
  • Cone to flask:
    24/29 NS