Biopsy processing/embedding cassettes, Swingsette®, M518
Grey
Catalog # 720-1683
Supplier: Simport Scientific
Biopsy processing/embedding cassettes, Swingsette®, M518
Catalog # 720-1683
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Ideal for cassette-labelling instruments since the covers are packed separately in the case. These biopsy cassettes, made of acetal polymer, are similar to the M517 series but are especially designed to hold biopsy specimens during the processing/embedding process as well as in storage cabinets.
- Moulded from a special high density polymer, they keep specimens safely submerged in liquid
- Totally resistant to the chemical action of histological solvents
- The efficient flow-through slots maximise fluid exchange and ensure proper drainage
- The special hinge make it easy to attach lid to base
This new model differs by the special hinge that holds the base and cover together. This hinge allows cassettes to be opened and closed as often as necessary. The cover can be removed and re-inserted easily without danger of specimen loss. Three frosted writing surfaces are made available, including the anterior one at a 45° angle. Available in several colours.
: Each case contains two dispenser boxes of 500 cassettes and one box of 1000 covers.
Specifications
- Colour:Grey