Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Swingsette®, M516
Purple
Catalog # 720-1642
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Swingsette®, M516
Catalog # 720-1642
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Swingsette®, M516
Biopsy processing/embedding cassettes, Swingsette®, M516
Purple
Price
315,00 €
Log in for availability
1 * 1.500 items
Product Details & Documents
Acetal polymer.
  • Integral hinged lid, with large tab for easier opening
  • Disposable plastic cassettes are highly resistant to histological solvents
  • Small, square openings ensure good fluid exchange without having to use foam pads or biopsy bags
  • Front face inclined 45° enabling use with certain automatic labelling systems
 : Each case contains three boxes of 500 cassettes.
Specifications
  • Colour:
    Purple