Orders LinkContactUsLinkComponent
Cover glasses, square
Cover glasses, square
24×24 mm, 0,13 - 0,16 mm, #1, Borosilicate glass
Catalog # HECH40990121
Cover glasses, square
Catalog # HECH40990121
Cover glasses, square
Cover glasses, square
24×24 mm, 0,13 - 0,16 mm, #1, Borosilicate glass
Price
32,70 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Borosilicate glass coverslips for histology and microscopy.
Specifications
  • No.:
    #1
  • L×W:
    24×24 mm
  • Material:
    Borosilicate glass
  • Thickness:
    0,13 - 0,16 mm