Order Entry
Export
Orders LinkContactUsLinkComponent
Personal eyewash

Personal eyewash

Holster for pocket model, green, with belt loop, 8×4,5×16 mm
Catalog # 111-0165
Supplier:   ORKLA CARE
Personal eyewash
Catalog # 111-0165
Supplier:  ORKLA CARE
Personal eyewash
Personal eyewash

Holster for pocket model, green, with belt loop, 8×4,5×16 mm

Price
13,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Sterile, buffered, isotonic sodium chloride solution for rinsing eyes following an accident. Available as a single-use, pocket version (235 ml) or wall mounted (500 ml). The bottle opens automatically when twisted out of the wall bracket, and must therefore, be used immediately.

  • Quick and easy to open
  • Eye cup protected from dust
  • Generous flow of fluid for effective rinsing - the eye cup on the bottle channels the fluid to the eye, avoiding unnecessary spills
  • Neutralises the pH value

Expiry date: 4 years if container not opened.

* Bottle holder is supplied without eyewash.

Documents

Specifications
  • Description:
    Holster for pocket model, green, with belt loop, 8×4,5×16 mm
  • Seq:
    05