Eppendorf® Protein LoBind Plates
Deepwell plate, White, 20 plates (5 bags × 4 plates), 1000 µl
Catalog # 737-2544
Supplier: EPPENDORF
Jump to:
Product Details & Documents
Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.
- Eppendorf LoBind material ensures excellent sample recovery for improved assay results
- Free of surface coating (e.g., silicone) to minimise the risk of sample interference
- Lot-certified PCR clean purity grade: Free of human DNA, DNase, RNase and PCR inhibitors
- High-contrast Unique OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors
- RecoverMax® well design: Optimised well geometry for minimal remaining/dead volume and excellent mixing properties
- Raised well rims and a smooth surface ensure reliable sealing in plates
Protein sample recovery can be maximised with Eppendorf Protein LoBind Plates. Eppendorf LoBind plates are designed with a special, two-component polymer mix, creating a hydrophilic surface to ensure optimal sample recovery rates. These plates are specially developed for protein research or sensitive proteomic test methods, providing superior results by reducing sample binding to the plate surface.
Purity grade: PCR clean
Material: Polypropylene (PP)
Autoclavable: 121 °C, 20 min
Centrifugation stability: 6000×g
Well colour: Clear
Specifications
- Packed:20 plates (5 bags × 4 plates)
- Colour:White
- Description:Deepwell plate
- Seq:05
- Well volume:1000 µl
- Cat. No.:737-2544
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact



