Spillage trays
Collecting tray, PEDH material, White, not conductive, without base insert, 300×400×170 mm, 235×335×160 mm
Catalog # 590-0809
Supplier: S.C.A.T.
Jump to:
Product Details & Documents
Made of HDPE, white or black (electrically conductive). Containers for all purposes.
- Electrically conductive available
- Available with base insert; the canisters always stay dry
- Anti-static mat and grund cable optionally available
Suitable for a wide range of canisters. Collect liquid in case of a leakage. The SCAT collection trays prevent the uncontrolled spread of hazardous liquids in the event of damage to a disposal container. The HDPE-EC conductive tray also prevents the build-up of static charge, which prevents the ignition of explosive substances.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Type:not conductive, without base insert
- W×D×H ext.:300×400×170 mm
- Colour:White
- Description:Collecting tray, PEDH material
- Seq:01
- W×D×H int.:235×335×160 mm



