Accessories for SCAT Level Controls
Extension cable for signal transmission, 10 m
Catalog # 590-0728
Supplier: S.C.A.T.
Jump to:
Product Details & Documents
Necessary and helpful accessories for use with SCAT electronic level controls.
- Switching cables
- Disk sensors
- Signal cables
- Power supply units
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Description:Extension cable for signal transmission, 10 m



