Order Entry
Export
Orders LinkContactUsLinkComponent
Containers, with screw cap, LaboPlast®

Containers, with screw cap, LaboPlast®

12 ml, 25 mm, 31 mm
Catalog # BURK6214-0012
Supplier:   BURKLE
Containers, with screw cap, LaboPlast®
Catalog # BURK6214-0012
Supplier:  BURKLE
Containers, with screw cap, LaboPlast®
Containers, with screw cap, LaboPlast®

12 ml, 25 mm, 31 mm

Price
0,30 €
Log in for availability
1 * 1 items
Product Details & Documents

PP, white.

  • Sterilisable
  • Material impermeable to light
  • Suitable for foodstguffs

Shapely packing container with screw cap. Emptying without residues because there are no crevices.

Specifications
  • Ext.Ø:
    25 mm
  • Capacity:
    12 ml
  • Height:
    31 mm