Ethanol absolute ≥99.5%, TechniSolv®, pure
Plastic container, Duty paid, 10 L
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Ethanol absolute GPR RECTAPUR Plastic bottle 25L 1 * 25 L Catalog #20820.464
Product 20820.464 is a replacement for 83813.410_T but varies by Brand, Packaging or Quantity.
Ethanol absolute TechniSolv pure 1 * 5 L Catalog #83813.360
Product 83813.360 is a replacement for 83813.410_T but varies by Brand, Packaging or Quantity.
Ethanol absolute TechniSolv pure 1 * 20 L Catalog #83813.440
Product 83813.440 is a replacement for 83813.410_T but varies by Brand, Packaging or Quantity.
Product Details & Documents
Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Merck Index:12,03806
- Boiling Pt:78.3 °C (1013 hPa)
- Melting Pt:–117 °C
- Density:0.7895 g/cm³ (20 °C)
- Flash Pt:12 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- REACH:01-2119457610-43
Documents
Certificates
Search by Lot Number10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
P20_VWRC83813.360_SPEC_EN.pdf P20_VWRC83813.410_SPEC_EN.pdf P20_VWRC83813.440_SPEC_EN.pdf spec_VWRC83813.360 spec_VWRC83813.410
Product specification
More
VWR Chemicals Big Packs and Bulks VWR Denatured Ethanol
Product Literature
Specifications
Specification Test Results
- Assay (V/V):Min. 99.5 %
- Density (20/20):0.788 - 0.794
- Water:Max. 0.5 %
Specifications
- Pack type:Plastic container
- Pk:10 L
- Duty status:Duty paid



