Ethanol ≥96% (v/v), TechniSolv®, pure
Plastic jerrican, non-removable head, Duty paid, 10 L
Ethanol ≥96% (v/v), TechniSolv®, pure
Catalog # 83804.410_T
CAS Number: 64-17-5
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Ethanol 96 % vol pure TechniSolv 1 * 5 L Catalog #83804.360
Product 83804.360 is a replacement for 83804.410_T but varies by Brand, Packaging or Quantity.
Ethanol 96 % vol pure TechniSolv 1 * 20 L Catalog #83804.440
Product 83804.440 is a replacement for 83804.410_T but varies by Brand, Packaging or Quantity.
Product Details & Documents
Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Boiling Pt:78.3 °C (1013 hPa)
- Melting Pt:–117 °C
- Density:0.7895 g/cm³ (20 °C)
- Flash Pt:12 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- REACH:01-2119457610-43
Certificates
Search by Lot Number10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Specifications
Specification Test Results
- Assay:Min. 95 %
- Assay (V/V):Min. 95 %
- Density (20/20):0.805 - 0.811
Specifications
- Pack type:Plastic jerrican, non-removable head
- Pk:10 L
- Duty status:Duty paid



