VWR®, Calibration Weights, Class 1
2000 g, 5000 ± mg
Catalog # 611-8764
Supplier: VWR Collection
Jump to:
Product Details & Documents
Stainless steel masses adjusted to class 1 for precise checking and adjustment of electronic balances.
: ASTM certified.
: Individual weights are supplied without certificate.
Documents
VWR® for Calibrations VWR® for Weighing
Product Literature
Specifications
- Weight:2000 g
- Seq:06
- Tolerance:5000 ± mg