Orders LinkContactUsLinkComponent
VWR®, Calibration Weights, Class 1
VWR®, Calibration Weights, Class 1
50 g, 120 ± mg
Catalog # 611-8759
Supplier:   VWR Collection
VWR®, Calibration Weights, Class 1
Catalog # 611-8759
Supplier:  VWR Collection
VWR®, Calibration Weights, Class 1
VWR®, Calibration Weights, Class 1
50 g, 120 ± mg
Price
87,80 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel masses adjusted to class 1 for precise checking and adjustment of electronic balances.
Certifications: ASTM certified.
Delivery information: Individual weights are supplied without certificate.
Documents
Specifications
  • Weight:
    50 g
  • Seq:
    01
  • Tolerance:
    120 ± mg